Anti-B3GAT1 Rabbit Polyclonal Antibody (DyLight® 650)
Lieferant: Novus Biologicals
NBP1-78982C
NOVUNBP1-78982CEA 722 EUR
NOVUNBP1-78982C
Anti-B3GAT1 Rabbit Polyclonal Antibody (DyLight® 650)
Antikörper
Type: Primary
Antigen: B3GAT1 (beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse