Anti-ZEB2 Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NBP1-82991
NOVUNBP1-82991EA 663 EUR
NOVUNBP1-82991
Anti-ZEB2 Rabbit Polyclonal Antibody
Antikörper
Analysis in human cell lines SK-MEL-30 and Caco-2 using Anti-ZEB2 antibody. Corresponding ZEB2 RNA-seq data are presented for the same cell lines. Loading control: Anti-HDAC1.
  • Immunohistochemistry 1:500 - 1:1000
  • Immunohistochemistry - Paraffin: 1:500 - 1:1000
  • Western blot: 0.04 to 0.4 ug/ml
Type: Primary
Antigen: ZEB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse