Anti-AAK1 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP1-84110
NOVUNBP1-84110EA 635 EUR
NOVUNBP1-84110
Anti-AAK1 Rabbit Polyclonal Antibody
Antikörper
The AAK1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to AAK1. This antibody reacts with human. The AAK1 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: AAK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR