Anti-Tight Junction Protein 1 Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NBP1-85046
NOVUNBP1-85046EA 663 EUR
NOVUNBP1-85046
Anti-Tight Junction Protein 1 Rabbit Polyclonal Antibody
Antikörper
The Tight Junction Protein 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Tight Junction Protein 1. This antibody reacts with human, mouse. The Tight Junction Protein 1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Tight Junction Protein 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse