Anti-FoxJ1/HFH4 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP1-87928
NOVUNBP1-87928EA 663 EUR
NOVUNBP1-87928
Anti-FoxJ1/HFH4 Rabbit Polyclonal Antibody
Antikörper
The FoxJ1 / HFH4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to FoxJ1 / HFH4. This antibody reacts with human. The FoxJ1 / HFH4 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: FoxJ1/HFH4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR