Anti-RNASET2 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP1-88753
NOVUNBP1-88753EA 635 EUR
NOVUNBP1-88753
Anti-RNASET2 Rabbit Polyclonal Antibody
Antikörper
The RNASET2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RNASET2. This antibody reacts with human. The RNASET2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: RNASET2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR