Anti-Ajuba Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NBP1-89570
NOVUNBP1-89570EA 652 EUR
NOVUNBP1-89570
Anti-Ajuba Rabbit Polyclonal Antibody
Antikörper
The Ajuba Antibody from Novus Biologicals is a rabbit polyclonal antibody to Ajuba. This antibody reacts with human. The Ajuba Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Ajuba
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human