Anti-Renin R Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP1-90820
NOVUNBP1-90820EA 644 EUR
NOVUNBP1-90820
Anti-Renin R Rabbit Polyclonal Antibody
Antikörper
The Renin R Antibody from Novus Biologicals is a rabbit polyclonal antibody to Renin R. This antibody reacts with human, mouse, rat. The Renin R Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Renin R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR