Anti-SETX Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: Novus Biologicals
NBP1-94712F
NOVUNBP1-94712FEA 722 EUR
NOVUNBP1-94712F
Anti-SETX Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Primary
Antigen: SETX (senataxin)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR