Human IgM Isotype Control (Biotin)

Lieferant: Novus Biologicals
NBP1-96989
NOVUNBP1-96989EA 363 EUR
NOVUNBP1-96989
Human IgM Isotype Control (Biotin)
Antikörper
Human IgM Protein [Biotin]. Tested Reactivity: Human.

Type: Control
Antigen: AGM1
Clonality:
Clone:
Conjugation: Biotin
Epitope:
Host: Human
Isotype: IgM
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR