Anti-SUV420H1 Rabbit Polyclonal Antibody (DyLight® 650)
Lieferant: Novus Biologicals
NBP1-97312C
NOVUNBP1-97312CEA 668 EUR
NOVUNBP1-97312C
Anti-SUV420H1 Rabbit Polyclonal Antibody (DyLight® 650)
Antikörper
Type: Primary
Antigen: SUV420H1 (suppressor of variegation 4-20 homolog 1 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse