Anti-LCMT1 Mouse Monoclonal Antibody [clone: 2C9]

Lieferant: Novus Biologicals
NBP2-00923
NOVUNBP2-00923EA 720 EUR
NOVUNBP2-00923
Anti-LCMT1 Mouse Monoclonal Antibody [clone: 2C9]
Antikörper
Mouse Monoclonal LCMT1 Antibody (2C9). Tested Applications: Western Blot, Immunohistochemistry. Tested Reactivity: Human, Mouse, Rat, Primate.

Type: Primary
Antigen: LCMT1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR