Anti-GBP1 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP2-03972
NOVUNBP2-03972EA 558 EUR
NOVUNBP2-03972
Anti-GBP1 Rabbit Polyclonal Antibody
Antikörper
The GBP1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to GBP1. This antibody reacts with human, mouse. The GBP1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 5 µg/ml

Type: Primary
Antigen: GBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR