Anti-SGK1 Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NBP2-19010
NOVUNBP2-19010EA 446 EUR
NOVUNBP2-19010
Anti-SGK1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SGK1 (serum/glucocorticoid regulated kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human