Anti-MKI67 Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NBP2-19012
NOVUNBP2-19012EA 450 EUR
NOVUNBP2-19012
Anti-MKI67 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MKI67 (marker of proliferation Ki-67)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human