Anti-PTGDR2 Mouse Monoclonal Antibody [clone: 54N1H4]

Lieferant: Novus Biologicals
NBP2-19013
NOVUNBP2-19013EA 558 EUR
NOVUNBP2-19013
Anti-PTGDR2 Mouse Monoclonal Antibody [clone: 54N1H4]
Antikörper
Type: Primary
Antigen: PTGDR2 (prostaglandin D2 receptor 2)
Clonality: monoclonal
Clone: 54N1H4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR