Anti-RUNX3 Mouse Monoclonal Antibody [clone: 2B10E8]

Lieferant: Novus Biologicals
NBP2-22391
NOVUNBP2-22391EA 558 EUR
NOVUNBP2-22391
Anti-RUNX3 Mouse Monoclonal Antibody [clone: 2B10E8]
Antikörper
Type: Primary
Antigen: RUNX3 (runt-related transcription factor 3)
Clonality: monoclonal
Clone: 2B10E8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR