Anti-ANXA1 Mouse Monoclonal Antibody [clone: 2F1]

Lieferant: Novus Biologicals
NBP2-23485SS NBP2-23485
NOVUNBP2-23485SSEA 262 EUR
NOVUNBP2-23485SS NOVUNBP2-23485
Anti-ANXA1 Mouse Monoclonal Antibody [clone: 2F1]
Antikörper
Type: Primary
Antigen: ANXA1 (annexin A1)
Clonality: monoclonal
Clone: 2F1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR