Anti-MAP3K14 Rabbit Polyclonal Antibody (DyLight® 650)
Lieferant: Novus Biologicals
NBP2-23603C
NOVUNBP2-23603CEA 668 EUR
NOVUNBP2-23603C
Anti-MAP3K14 Rabbit Polyclonal Antibody (DyLight® 650)
Antikörper
Type: Primary
Antigen: MAP3K14 (mitogen-activated protein kinase kinase kinase 14)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse