Anti-MAP3K14 Rabbit Polyclonal Antibody (DyLight® 350)

Lieferant: Novus Biologicals
NBP2-23603UV
NOVUNBP2-23603UVEA 668 EUR
NOVUNBP2-23603UV
Anti-MAP3K14 Rabbit Polyclonal Antibody (DyLight® 350)
Antikörper
Type: Primary
Antigen: MAP3K14 (mitogen-activated protein kinase kinase kinase 14)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 350
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR