Anti-ROR gamma Mouse Monoclonal Antibody [clone: 4G419]

Lieferant: Novus Biologicals
NBP2-24449
NOVUNBP2-24449EA 596 EUR
NOVUNBP2-24449
Anti-ROR gamma Mouse Monoclonal Antibody [clone: 4G419]
Antikörper
Type: Primary
Antigen: ROR gamma
Clonality: monoclonal
Clone: 4G419
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR