Anti-IGHG1 Rabbit Polyclonal Antibody (PerCP)

Lieferant: Novus Biologicals
NBP2-24891PCP
NOVUNBP2-24891PCPEA 0 EUR
NOVUNBP2-24891PCP
Anti-IGHG1 Rabbit Polyclonal Antibody (PerCP)
Antikörper
Type:
Antigen: IGHG1
Clonality: Polyclonal
Clone:
Conjugation: PerCP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR