Rabbit IgG Isotype Control (FITC (Fluorescein Isothiocyanate))
Lieferant: Novus Biologicals
NBP2-24892
NOVUNBP2-24892EA 571 EUR
NOVUNBP2-24892
Rabbit IgG Isotype Control (FITC (Fluorescein Isothiocyanate))
Antikörper
Type: Control
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rabbit