Anti-CA15-3 Mouse Monoclonal Antibody [clone: DBMM1]
Lieferant: Novus Biologicals
NBP2-26672
NOVUNBP2-26672EA 835 EUR
NOVUNBP2-26672
Anti-CA15-3 Mouse Monoclonal Antibody [clone: DBMM1]
Antikörper
The CA15-3 Antibody (DBMM1) from Novus Biologicals is a mouse monoclonal antibody to CA15-3. This antibody reacts with human. The CA15-3 Antibody (DBMM1) has been validated for the following applications: Immunohistochemistry-paraffin.
Type: Primary
Antigen: CA15-3
Clonality: Monoclonal
Clone: DBMM1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human