Anti-IFI16 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP2-27066
NOVUNBP2-27066EA 572 EUR
NOVUNBP2-27066
Anti-IFI16 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IFI16 (interferon, gamma-inducible protein 16)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR