Anti-SIGLEC9 Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NBP2-27070
NOVUNBP2-27070EA 434 EUR
NOVUNBP2-27070
Anti-SIGLEC9 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SIGLEC9 (sialic acid binding Ig-like lectin 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen