Anti-EBI3 Mouse Monoclonal Antibody [clone: 42N1G4]
Lieferant: Novus Biologicals
NBP2-27356
NOVUNBP2-27356EA 558 EUR
NOVUNBP2-27356
Anti-EBI3 Mouse Monoclonal Antibody [clone: 42N1G4]
Antikörper
Type: Primary
Antigen: EBI3 (Epstein-Barr virus induced 3)
Clonality: monoclonal
Clone: 42N1G4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human