Anti-IL17RE Mouse Monoclonal Antibody [clone: 46N7E3]
Lieferant: Novus Biologicals
NBP2-27367
NOVUNBP2-27367EA 596 EUR
NOVUNBP2-27367
Anti-IL17RE Mouse Monoclonal Antibody [clone: 46N7E3]
Antikörper
Type: Primary
Antigen: IL17RE (interleukin 17 receptor E)
Clonality: monoclonal
Clone: 46N7E3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human