Anti-LIF Rat Monoclonal Antibody [clone: 39N7D10]

Lieferant: Novus Biologicals
NBP2-27406
NOVUNBP2-27406EA 572 EUR
NOVUNBP2-27406
Anti-LIF Rat Monoclonal Antibody [clone: 39N7D10]
Antikörper
Type: Primary
Antigen: LIF (leukemia inhibitory factor)
Clonality: monoclonal
Clone: 39N7D10
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR