Anti-H2AFX Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NBP2-27428
NOVUNBP2-27428EA 444 EUR
NOVUNBP2-27428
Anti-H2AFX Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: H2AFX (H2A histone family, member X)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human