Anti-MATH2/NEUROD6 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP2298650.025
NOVUNBP2298650.025EA 0 EUR
NOVUNBP2298650.025
Anti-MATH2/NEUROD6 Rabbit Polyclonal Antibody
Antikörper
Bitte beachten This product is for research use only and is not approved for use in humans or in clinical diagnosis.

Type: Primary
Antigen: MATH2/NEUROD6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR