Anti-Nectin-2/CD112 Rabbit Monoclonal Antibody [clone: S06-8F3]
Lieferant: Novus Biologicals
NBP3-20070-50
NOVUNBP3-20070-50EA 536 EUR
NOVUNBP3-20070-50
Anti-Nectin-2/CD112 Rabbit Monoclonal Antibody [clone: S06-8F3]
Antikörper
Nectin 2 encodes a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions.
  • Formulation: 50 mM tris-glycine (pH 7,4), 0,15 M NaCl, 40% glycerol, 0,05% BSA
  • Preservative: 0,01% sodium azide
It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterised.
Type: Recombinant
Antigen: Nectin-2/CD112
Clonality: Monoclonal
Clone: S06-8F3
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat