Anti-BLOS3 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP303343-100
NOVUNBP303343-100EA 592 EUR
NOVUNBP303343-100
Anti-BLOS3 Rabbit Polyclonal Antibody
Antikörper
Immunocytochemistry/ Immunofluorescence 1:50 - 1:200; Immunohistochemistry 1:50 - 1:200; Immunohistochemistry-Paraffin.

Type: Primary
Antigen: BLOS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR