Anti-GLP1R Rabbit Polyclonal Antibody (DyLight® 680)
Lieferant: Novus Biologicals
NLS1205FR
NOVUNLS1205FREA 690 EUR
NOVUNLS1205FR
Anti-GLP1R Rabbit Polyclonal Antibody (DyLight® 680)
Antikörper
Type: Primary
Antigen: GLP1R (glucagon-like peptide 1 receptor)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 680
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse