Anti-OPRD1 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NLS222
NOVUNLS222EA 655 EUR
NOVUNLS222
Anti-OPRD1 Rabbit Polyclonal Antibody
Antikörper
The delta Opioid R / OPRD1 antibody from Novus Biologicals is a rabbit polyclonal antibody to delta Opioid R / OPRD1. This antibody reacts with human. The delta Opioid R / OPRD1 antibody has been validated for the following applications: immunohistochemistry, immunohistochemistry-paraffin.

Type: Primary
Antigen: OPRD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR