Anti-GPR119 Rabbit Polyclonal Antibody (Biotin)

Lieferant: Novus Biologicals
NLS548B
NOVUNLS548BEA 686 EUR
NOVUNLS548B
Anti-GPR119 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: GPR119 (G protein-coupled receptor 119)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR