Anti-Pan-Ras (Ab-1) (F132-62) Mouse Monoclonal Antibody [clone: F132-62]
Lieferant: Merck Millipore (Oncogene)
OP21-100
OP21-100EA 583 EUR
OP21-100
Anti-Pan-Ras (Ab-1) (F132-62) Mouse Monoclonal Antibody [clone: F132-62]
Antikörper
Type: Primary
Antigen: Pan-Ras (Ab-1) (F132-62)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen