Anti-PECAM1 Mouse Monoclonal Antibody [clone: 8A9]

Lieferant: OriGene
CF500121
ORIGCF500121EA 833 EUR
ORIGCF500121
Anti-PECAM1 Mouse Monoclonal Antibody [clone: 8A9]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50

Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone: 8A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR