Anti-VEGF Mouse Monoclonal Antibody [clone: 4E3]

Lieferant: OriGene
CF500289
ORIGCF500289EA 833 EUR
ORIGCF500289
Anti-VEGF Mouse Monoclonal Antibody [clone: 4E3]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: VEGF
Clonality: Monoclonal
Clone: 4E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR