Anti-GBP2 Mouse Monoclonal Antibody [clone: 2G6]

Lieferant: OriGene
CF500655
ORIGCF500655EA 833 EUR
ORIGCF500655
Anti-GBP2 Mouse Monoclonal Antibody [clone: 2G6]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: GBP2
Clonality: Monoclonal
Clone: 2G6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR