Anti-GPI Mouse Monoclonal Antibody [clone: 5G9]

Lieferant: OriGene
CF501127
ORIGCF501127EA 833 EUR
ORIGCF501127
Anti-GPI Mouse Monoclonal Antibody [clone: 5G9]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: GPI
Clonality: Monoclonal
Clone: 5G9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR