Anti-MPP3 Mouse Monoclonal Antibody [clone: 5E8]

Lieferant: OriGene
CF501304
ORIGCF501304EA 833 EUR
ORIGCF501304
Anti-MPP3 Mouse Monoclonal Antibody [clone: 5E8]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: MPP3
Clonality: Monoclonal
Clone: 5E8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR