Anti-IGF2BP2 Mouse Monoclonal Antibody [clone: 4F7]

Lieferant: OriGene
CF501315
ORIGCF501315EA 833 EUR
ORIGCF501315
Anti-IGF2BP2 Mouse Monoclonal Antibody [clone: 4F7]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: IGF2BP2
Clonality: Monoclonal
Clone: 4F7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR