Anti-HIBCH Mouse Monoclonal Antibody [clone: 2A5]

Lieferant: OriGene
CF501318
ORIGCF501318EA 833 EUR
ORIGCF501318
Anti-HIBCH Mouse Monoclonal Antibody [clone: 2A5]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HIBCH
Clonality: Monoclonal
Clone: 2A5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR