Anti-MAPK7 Mouse Monoclonal Antibody [clone: 3H2]

Lieferant: OriGene
CF502119
ORIGCF502119EA 833 EUR
ORIGCF502119
Anti-MAPK7 Mouse Monoclonal Antibody [clone: 3H2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MAPK7
Clonality: Monoclonal
Clone: 3H2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR