Anti-ACY3 Mouse Monoclonal Antibody [clone: 3A1]

Lieferant: OriGene
CF502292
ORIGCF502292EA 833 EUR
ORIGCF502292
Anti-ACY3 Mouse Monoclonal Antibody [clone: 3A1]
Antikörper
Recommended Dilutions: Flow Cytometry: 1:100

Type: Primary
Antigen: ACY3
Clonality: Monoclonal
Clone: 3A1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR