Anti-STAT5A Mouse Monoclonal Antibody [clone: 10H1]

Lieferant: OriGene
CF502735
ORIGCF502735EA 833 EUR
ORIGCF502735
Anti-STAT5A Mouse Monoclonal Antibody [clone: 10H1]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: STAT5A
Clonality: Monoclonal
Clone: 10H1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR