Anti-STAT5A Mouse Monoclonal Antibody [clone: 4E7]

Lieferant: OriGene
CF502738
ORIGCF502738EA 833 EUR
ORIGCF502738
Anti-STAT5A Mouse Monoclonal Antibody [clone: 4E7]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: STAT5A
Clonality: Monoclonal
Clone: 4E7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR