Anti-NDUFA7 Mouse Monoclonal Antibody [clone: 2C4]

Lieferant: OriGene
CF502749
ORIGCF502749EA 833 EUR
ORIGCF502749
Anti-NDUFA7 Mouse Monoclonal Antibody [clone: 2C4]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: NDUFA7
Clonality: Monoclonal
Clone: 2C4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR