Anti-TP53 Mouse Monoclonal Antibody [clone: 2E4]

Lieferant: OriGene
CF502783
ORIGCF502783EA 833 EUR
ORIGCF502783
Anti-TP53 Mouse Monoclonal Antibody [clone: 2E4]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: 2E4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR